Anti-RUFY1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIR9907-01EA 551 EUR
USBIR9907-01
Anti-RUFY1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 05-0.2?g/ml

Type: Primary
Antigen: RUFY1 (RUN and FYVE domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR