Anti-S100A4 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIS0051-83BEA 441 EUR
USBIS0051-83B
Anti-S100A4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:100-1:200

Type: Primary
Antigen: S100A4 (S100 calcium binding protein A4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Cat, Dog, Horse, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR