Anti-SATB1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIS0098-85CEA 551 EUR
USBIS0098-85C
Anti-SATB1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: SATB1 (SATB homeobox 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR