Anti-SIAH1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIS1013-42EA 551 EUR
USBIS1013-42
Anti-SIAH1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: SIAH1 (siah E3 ubiquitin protein ligase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR