Anti-Smallpox A27L Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIS1014-58U1EA 507 EUR
USBIS1014-58U1
Anti-Smallpox A27L Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Smallpox A27L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR