Anti-SOD1 Mouse Monoclonal Antibody [clone: 2F5]

Lieferant: United States Biological
USBIS8060-33CEA 633 EUR
USBIS8060-33C
Anti-SOD1 Mouse Monoclonal Antibody [clone: 2F5]
Antikörper
Recommended dilution: Immunochemistry: 15 ?g/ml

Type: Primary
Antigen: SOD1 (superoxide dismutase 1, soluble)
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR