Anti-TNS1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIT2600-15BEA 551 EUR
USBIT2600-15B
Anti-TNS1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:2000; Western Blot: 1 ?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: TNS1 (tensin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR