Anti-TLR3 Mouse Monoclonal Antibody [clone: TLR3.7]

Lieferant: United States Biological
USBIT8050-20REA 802 EUR
USBIT8050-20R
Anti-TLR3 Mouse Monoclonal Antibody [clone: TLR3.7]
Antikörper
Recommended dilution: Flow Cytometry: 1:50

Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: monoclonal
Clone: TLR3.7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR