Smooth muscle cell growth medium 2
Lieferant: PROMOCELL
C-22062 C-22162 C-22267 C-22262
C-22062EA 274 EUR
C-22062 C-22162 C-22267 C-22262
Smooth muscle cell growth medium 2
Zellkulturmedien
Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium designed to establish and maintain smooth muscle cell cultures.
The medium is optimised for primary human cells but can also be used for bovine, porcine, murine, dog and rat smooth muscle cells.
Lieferumfang: PromoCell Smooth Muscle Cell Growth Medium 3 is available as Medium (ready to use) or as Medium Kit. The Medium (ready to use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. The Medium Kit consists of a 500 ml bottle of Basal Medium and the SupplementPack (a set of individual vials with pre-measured supplements) allowing the user full control over the media formulation. Adding the SupplementMix or the SupplementPack to the Basal Medium results in the complete Growth Medium. Basal Medium (with or without phenol red) as well as the SupplementMix and SupplementPack can also be purchased separately.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable