Adipocyte nutrition medium
Lieferant: PROMOCELL
C-27438
C-27439
C-27431
C-27435
C-27438EA
228
EUR
C-27438
C-27439
C-27431
C-27435
Adipocyte nutrition medium
Zellkulturmedien
PromoCell Adipocyte Nutrition Medium is designed for the cultivation of mature adipocytes. The medium is optimised for primary human cells but can also be used for murine adipocytes.
Lieferumfang: The medium consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. Medium kit consists of 1× 500 ml bottle of basal medium and 1× SupplementPack (individual vials with pre-measured supplements).