Fibroblast culture medium
Lieferant: PROMOCELL
C-23210 C-23010 C-23110 C-23215
C-23210EA 196 EUR
C-23210 C-23010 C-23110 C-23215
Fibroblast culture medium
Zellkulturmedien
PromoCell Fibroblast Culture Medium is a serum-free medium optimised for the cultivation of neonatal and juvenile fibroblasts.
The medium is optimised for primary human cells but can also be used for rat, porcine and bovine fibroblasts as well as fibroblast cell lines.
Lieferumfang: The medium (ready-to-use) consists of 1× 500 ml bottle of basal medium and 1× vial of SupplementMix. Medium Kit consists of 1× 500 ml bottle basal medium and 1× SupplementPack (individual vials with pre-measured supplements).