Human preadipocytes
Lieferant: PROMOCELL
C-14072 C-14073
C-14072EA 249 EUR
C-14072 C-14073
Human preadipocytes
Zellen
Primary Human White Preadipocytes (HWP) are isolated from adult subcutaneous or visceral adipose tissue from different locations (lot specific source information is available on request). Adipose tissue is crucial in energy storage and metabolic homeostasis. An increase in adipose tissue results either from an enlargement of mature adipocytes or from the differentiation of adipocyte precursor cells (preadipocytes) into new mature adipocytes. These preadipocytes are already present in adipose tissues. They can proliferate throughout adult life and replace the cells that have differentiated into mature adipocytes.
Since this differentiation process is controlled by a variety of growth factors and hormones, preadipocytes are suitable for the investigation of the physiological mechanisms controlling proliferation, differentiation, and function of adipose tissue.
The PromoCell adipocyte cell culture system provides preadipocytes with optimized growth media and serum-free differentiation media.
Bestellinformation: Primary Human White Preadipocytes can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.