Human follicle dermal papilla cells
Lieferant: PROMOCELL
Primary human follicle dermal papilla cells (HFDPC) are isolated from human dermis of the scalp, originating from the occipital or temple region. HFDPC stain positive for alkaline phosphatase.
Dermal papillae are embedded in a laminin and collagen IV rich extracellular matrix at the base of the hair follicles. They are essential for the induction and maintenance of hair growth. Since they have androgen receptors, they can be used for in vitro screening of androgen blocking reagents.
Applications include: Hair growth research, drug testing, toxicological testing.