Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 488)

Lieferant: Agrisera
AGRIAS09631EA 218 EUR
AGRIAS09631
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR