Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Agrisera
AGRIAS10659EA 218 EUR
AGRIAS10659
Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Goat, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR