Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)

Lieferant: Agrisera
AGRIAS10661EA 246 EUR
AGRIAS10661
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR