Anti-IgM µ Goat Polyclonal Antibody (DyLight® 550)

Lieferant: Agrisera
AGRIAS121919EA 218 EUR
AGRIAS121919
Anti-IgM µ Goat Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR