Anti-Heat Shock Protein 90 Mouse Monoclonal Antibody [clone: 7G31]

Lieferant: United States Biological
USBIH1831-58-100EA 377 EUR
USBIH1831-58-100 USBIH1831-58-50
Anti-Heat Shock Protein 90 Mouse Monoclonal Antibody [clone: 7G31]
Antikörper
Recommended dilution: Western Blot: 1:500-1:2000; Immunochemistry: 1:100-1:200

Type: Primary
Antigen: Heat Shock Protein 90
Clonality: Monoclonal
Clone: 7G31
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR