Anti-Insulin-Like Growth Factor Binding Protein 7 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBII7661-17A-100EA 425 EUR
USBII7661-17A-100
Anti-Insulin-Like Growth Factor Binding Protein 7 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-2?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Insulin-Like Growth Factor Binding Protein 7
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR