Anti-Granulocyte Macrophage Colony Stimulating Factor Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIG8951-32-20EA 337 EUR
USBIG8951-32-20 USBIG8951-32-100
Anti-Granulocyte Macrophage Colony Stimulating Factor Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Granulocyte Macrophage Colony Stimulating Factor
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR