Anti-pro-Guanylin Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIG9805-04C-100EA 741 EUR
USBIG9805-04C-100 USBIG9805-04C-20
Anti-pro-Guanylin Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: pro-Guanylin
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR