Anti-Heat Shock Protein 27 Mouse Monoclonal Antibody [clone: 10F714]

Lieferant: United States Biological
USBIH1830-53G-25EA 349 EUR
USBIH1830-53G-25 USBIH1830-53G-100
Anti-Heat Shock Protein 27 Mouse Monoclonal Antibody [clone: 10F714]
Antikörper
Recommended dilution: Immunofluorescence:1:200; Western Blot:1:5000

Type: Primary
Antigen: Heat Shock Protein 27
Clonality: Monoclonal
Clone: 10F714
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Guinea pig, Hamster, Human, Rat,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR