Anti-IGF1R Rabbit Polyclonal Antibody
Lieferant: United States Biological
USBII7661-18U-100EA 481 EUR
USBII7661-18U-100
Anti-IGF1R Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000
Type: Primary
Antigen: IGF1R
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat,