Anti-CD31 Rat Monoclonal Antibody [clone: 390]

Lieferant: United States Biological
USBIC2383-20A-500EA 533 EUR
USBIC2383-20A-500
Anti-CD31 Rat Monoclonal Antibody [clone: 390]
Antikörper
Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 390
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR