Anti-Y Rabbit Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
ROCK600-406-262EA 814 EUR
ROCK600-406-262
Anti-Y Rabbit Polyclonal Antibody (Biotin)
Antikörper
Anti-PhosphoTyrosine antibody is suitable for use in ELISA and western blotting assays.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: Y
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR