Anti-Theophylline Goat Polyclonal Antibody

Lieferant: Fitzgerald Industries
FITZ20-TG158EA 0 EUR
FITZ20-TG158
Anti-Theophylline Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Theophylline
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR