Anti-ARL2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP16648CEA 348 EUR
ABGEAP16648C
Anti-ARL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR