Anti-MBP Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB7988EA 518 EUR
ABNOPAB7988
Anti-MBP Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of MBP.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR