IgG Isotype Control (AP (Alkaline Phosphatase))

Lieferant: Abnova
ABNOPAB19458EA 399 EUR
ABNOPAB19458
IgG Isotype Control (AP (Alkaline Phosphatase))
Antikörper
Mouse IgG raised from normal mouse serum.

Recommended Dilutions: Flow Cytometry: <1 ?g/106 cells; ELISA: 1:500-1:5000

Type: Control
Antigen: IgG
Clonality: AP (Alkaline Pohsphatase)
Clone: Monoclonal
Conjugation:
Epitope:
Host: IgG
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR