Anti-COL3A1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB13501EA 643 EUR
ABNOPAB13501
Anti-COL3A1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against native Col3a1.

Recommended Dilutions: ELISA: 1:200; Immunohistochemistry: 1:500; Immunofluorescence: 1:80

Type: Primary
Antigen: COL3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR