Anti-IL7 Rat Monoclonal Antibody [clone: BVD10-40F6]
Lieferant: Abnova
ABNOMAB3298EA 518 EUR
ABNOMAB3298
Anti-IL7 Rat Monoclonal Antibody [clone: BVD10-40F6]
Antikörper
Rat monoclonal antibody raised against recombinant IL7.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: IL7
Clonality: Monoclonal
Clone: BVD10-40F6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human