Anti-CCND1 Mouse Monoclonal Antibody [clone: CY-D1]

Lieferant: Abnova
ABNOMAB1449EA 518 EUR
ABNOMAB1449
Anti-CCND1 Mouse Monoclonal Antibody [clone: CY-D1]
Antikörper
Mouse monoclonal antibody raised against full length recombinant CCND1.

Recommended Dilutions: Western Blot: 4 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 8 ?g/ml; Immunohistochemistry (Frozen sections): 8 ?g/ml

Type: Primary
Antigen: CCND1
Clonality: Monoclonal
Clone: CY-D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR