Anti-APP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abnova
ABNOPAB28937EA 643 EUR
ABNOPAB28937
Anti-APP Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Rabbit polyclonal antibody against amyloid oligomers.

Recommended Dilutions: Immunohistochemistry: 1:1000-10000; Immunoprecipitation: 1:1000; Western Blot

Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR