Anti-beta NGF Chicken Polyclonal Antibody

Lieferant: Biosensis
BSENC-1558-500EA 312 EUR
BSENC-1558-500
Anti-beta NGF Chicken Polyclonal Antibody
Antikörper
FUNCTION: Nerve growth factor is important for the development and maintenance of the sympathetic and sensory nervous systems. It stimulates division and differentiation of sympathetic and embryonic sensory neurons. SUBUNIT: Homodimer, associated by noncovalent forces. SUBCELLULAR LOCATION: Secreted protein. SIMILARITY: Belongs to the NGF-beta family.

Antibody recognizes NGF in human, mouse and rat and likely other species as well as NGF is highly conserved but they have not been tested. Approved applications include: ELISA, WB and IHC (frozen, non-paraffin embedded tissues tested); not yet tested in paraffin tissues. WB suggested dilution of 1:500-1:2,000. IHC suggested dilution of 1:50-1:1,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Application Information:
ELISA, WB and IHC. WB suggested dilution of 1:500-1:2,000. IHC suggested dilution of 1:50-1:1,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: beta NGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR