Transport swabs
Lieferant: THERMO FISHER DIAGNOSTICS
TS0001A TS0002A
OXOITS0001AEA 457 EUR
OXOITS0001A OXOITS0002A
Transport swabs
Tupfer Transport-Tupfer
Effective transportation of throat, vaginal, wound and skin swabs conforming to highest medical device classification - Class II a surgically invasive transient use. All swabs have soft rayon tips, which are inert and non-toxic to micro-organisms.
The addition of charcoal in the medium is considered to neutralise bacterial toxins and other inhibitory substances and has been shown to increase the recovery of Neisseria gonorrhoeae.
  • Prevents disintegration of gel column during transport
  • Eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904