HPLC columns, Allure® Basix (USP L10)
Lieferant: Restek
9161555 9161562 9161565 9161575
REST9161555EA 0 EUR
REST9161555 REST9161562 REST9161565 REST9161575
HPLC columns, Allure® Basix (USP L10)
Chromatographiesäulen
Highly retentive propyl cyano phase. Excellent choice for basic compounds and for analytes containing amine group functionality.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Kohlenstoffbelastung 12%
Endcapped Ja
pH-Bereich 2,5 - 7,5
Temperaturlimit 80 °C
USP-Code L10