(1S)-(-)-3-Oxocamphorsulphonylimine zur Synthese
Lieferant: Merck
Synonyme:
3H-3a,6-Methano-2,1-benzisothiazol-7(4H)-one, 5,6-dihydro-8,8-dimethyl-, 2,2-dioxide, (3aS,6S)-, (1S,7S)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.01,5]dec-4-en-6-one 3,3-dioxide
M-Clarity™ Qualitätsstufe = MQ200
8.14388.0001
8.14388.0005
8.14388.0001EA
0
EUR
8.14388.0001
8.14388.0005
(1S)-(-)-3-Oxocamphorsulphonylimine zur Synthese
(1S)-(-)-3-Oxocamphorsulphonylimine
Formel:
C₁₀H₁₃NO₃S Molecular Weight: 227,28 g/mol |
MDL:
MFCD00066268 CAS-Nummer: 119106-38-6 |
Spezifikation Testergebnisse
Solubility | Insoluble (20 °C) |
Melting Point | 195 - 198 °C |
Molar Mass | 227.28 g/mol |
Hill Formula | C10H13NO3S |
Assay (of S) | ≥ 98% (m) |
Specific rotation | -192 to -190° |
Storage temperature | +15...+25 °C |
Identity (IR) | Conforms |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...