Chiral columns, Ultron ES
Lieferant: Agilent
702111610 702111651 702111651A 712111630 722111723 724111653 822111631A 822111651 832111630 902111651
HEWL702111610EA 3258 EUR
HEWL702111610 HEWL702111651 HEWL702111651A HEWL712111630 HEWL722111723 HEWL724111653 HEWL822111631A HEWL822111651 HEWL832111630 HEWL902111651
Chiral columns, Ultron ES
Chromatographiesäulen
Ultron chiral columns allow for direct racemic separations without derivatisation.
  • Direct racemic separations without derivatisation
Ultron ES-OVM chiral columns are used for the separation enantiomers of pharmaceuticals, such as hexobarbital, ibuprofen, and profenamine.
Ultron ES-Pepsin chiral columns are best suited to separate basic compounds that are difficult to separate by other columns.
ES-OVM contains an ovomucoid protein while ES-Pepsin columns have pepsin bonded to 120 Å, 5 µm silica particles
Both types of chiral columns are usable with mobile phases such as acetonitrile or ethanol and phosphate buffer.
Ultron ES-OVM-C is a column used for the separation of chiral isomers (enantiomeric compounds). This column recognises hydrogen-bonding, polar, ionic, and hydrophobic sites, as well as the three-dimensional structure of sample molecules. These particles are silica-based, nominally 5 µm in diameter with 120 Å pores.