Phenylfluorone zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 9-Phenyl-2,3,7-trihydroxy-6-fluorone, 9-Phenyl-3-fluorone, 2,6,7-Trihydroxy-9-phenyl-3-isoxanthone

M-Clarity™ Qualitätsstufe = MQ200

8.41569.0001 8.41569.0005
8.41569.0001EA 32.5 EUR
8.41569.0001 8.41569.0005
Phenylfluorone zur Synthese, Sigma-Aldrich®
Phenylfluorone
Formel: C₁₉H₁₂O₅
Molecular Weight: 320,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005048
CAS-Nummer: 975-17-7
EINECS: 213-550-3

Bestellen

Spezifikation Testergebnisse

Solubility 300 °C
Melting Point 300 °C
Hill Formula C19H12O5
Molar Mass 320,3 g/mol
HS Code 2932 99 00
Storage temperature 15...25 °C
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR