Anti-NAALADL1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP18189BEA 348 EUR
ABGEAP18189B
Anti-NAALADL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAALADL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR