Anti-CTSD Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP20675BEA 348 EUR
ABGEAP20675B
Anti-CTSD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CTSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR