Anti-ESR2 Mouse Monoclonal Antibody [clone: 555CT18.5.5]

Lieferant: Abgent
ABGEAM2112BEA 348 EUR
ABGEAM2112B
Anti-ESR2 Mouse Monoclonal Antibody [clone: 555CT18.5.5]
Antikörper
Type: Primary
Antigen: ESR2
Clonality: Monoclonal
Clone: 555CT18.5.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR