Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I) (21% Rh)

Lieferant: Thermo Fisher Scientific

Synonyme: Acetylacetonatocarbonyl triphenylphosphine Rhodium(I)

10458.01 10458.03 10458.06
10458.01EA 132 EUR
10458.01 10458.03 10458.06
Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I) (21% Rh)
Carbonyl-2,4-pentanedionato(triphenylphosphine)rhodium(I)

CAS No.: 25470-96-6 Molecular Formula: C24H22O3PRh Formula Weight: 492.32 Physical Form: Crystals Appearance: Yellow Solubility: Soluble in acetone and chlorinated solvents MDL No.: MFCD00064611


Applications: Hydroformylation

Formel: C₂₄H₂₂O₃PRh
Molecular Weight: 492.32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064611
CAS-Nummer: 25470-96-6
EINECS: 247-015-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR