trans-Dichlorobis(diethylsulphide)platinum(II) (43,7% Pt)

Lieferant: Thermo Scientific
Achtung

10484.03 10484.06
10484.03EA 128 EUR
10484.03 10484.06
trans-Dichlorobis(diethylsulphide)platinum(II) (43,7% Pt)
trans-Dichlorobis(diethylsulphide)platinum(II)

CAS No.: 15337-84-5 Molecular Formula: Pt[(C2H5)2S]2Cl2 Formula Weight: 446.37 Physical Form: Crystalline Solubility: Soluble in acetone, alcohol and benzene MDL No.: MFCD00050943


Applications: Hydrosilation

Formel: Pt[(C₂H₅)₂S]₂Cl₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00050943
CAS-Nummer: 15337-84-5
EINECS: 239-373-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR