Dichloro-(1,5-cyclooctadien)-palladium(II) (36,7% Pd)
Lieferant: Thermo Scientific
Achtung
Synonyme: 1,5-Cyclooctadienepalladium(II) Dichloride
10493.03 10493.06
10493.03EA 123 EUR
10493.03 10493.06
Dichloro-(1,5-cyclooctadien)-palladium(II) (36,7% Pd)
Dichloro(1,5-cyclooctadiene)palladium(II)
CAS No.: 12107-56-1 Molecular Formula: C8H12Cl2Pd Formula Weight: 285.49 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Yellow Solubility: Soluble in dichloromethane MDL No.: MFCD00012412
Formel: C₈H₁₂Cl₂Pd
Molecular Weight: 285,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012412
CAS-Nummer: 12107-56-1
EINECS: 235-161-8