Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer
Lieferant: Thermo Scientific
Synonyme:
Dichloro(cycloocta-1,5-diene)ruthenium(II), Dichloro(1,5-cyclooctadiene)ruthenium(II) polymer
10494.03
10494.06
10494.03EA
111
EUR
10494.03
10494.06
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer
CAS No.: 50982-12-2
Molecular Formula: [RuCl2(C8H12)]n
Physical Form: Powder
Appearance: Brown to black
Melting Point: 200° dec.
MDL No.: MFCD00171304
Formel:
[RuCl₂(C₈H₁₂)]n Molecular Weight: 280,1569 g/mol Schmelzpunkt: 200 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00171304 CAS-Nummer: 50982-12-2 |