Kaliumtetracyanoplatinat(II) Trihydrat (≥44,9% Pt) ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
10536.03
10536.06
10536.03EA
173
EUR
10536.03
10536.06
Kaliumtetracyanoplatinat(II) Trihydrat (≥44,9% Pt) ≥99,9% (Metall-Basis)
Kaliumtetracyanoplatinat(II) Trihydrat
Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.
Soluble in hot water
Formel:
C₄K₂N₄Pt·3H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149921 CAS-Nummer: 38245-27-1 EINECS: 209-236-0 UN: 1588 ADR: 6.1,III Merck Index: 14,7685 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...