Ammonium bis(oxalato)oxotitanate(IV) hydrate ≥99,998% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific
Achtung

10895.09 10895.18
10895.09EA 84.3 EUR
10895.09 10895.18
Ammonium bis(oxalato)oxotitanate(IV) hydrate ≥99,998% (Metall-Basis), Puratronic®
Ammonium bis(oxalato)oxotitanate(IV) hydrate

Purity: 99.998% CAS No.: 10580-03-7 Molecular Formula: (NH4)2TiO(C2O4)2.xH2O (x≈1) Formula Weight: 276.01(anhy) Physical Form: Crystalline powder Odour: Odourless UN No.: UN3288 MDL No.: MFCD00149620

Formel: (NH₄)₂TiO(C₂O₄)₂·xH₂O (x≈1)
Molecular Weight: 276,01 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149620
CAS-Nummer: 10580-03-7
EINECS: 234-174-6
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR