Tetraamminepalladium(II) chloride monohydrate (≥39% Pd) ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Tetraamminepalladium(II) chloride hydrate
11036.04
11036.09
11036.04EA
284
EUR
11036.04
11036.09
Tetraamminepalladium(II) chloride monohydrate (≥39% Pd) ≥99,9% (Metall-Basis)
Tetraamminepalladium(II) chloride monohydrate
Purity: 99.9%
CAS No.: 13933-31-8
Molecular Formula: Pd(NH3)4Cl2.H2O
Formula Weight: 263.45 (245.43anhy)
Physical Form: Crystalline
Density: 1.91
Sensitivity: Hygroscopic
Appearance: Colourless to yellow
Melting Point: 120° dec.
UN No.: UN3077
MDL No.: MFCD00151033
Formel:
Cl₂H₁₄N₄OPd Molecular Weight: 263,459 g/mol Schmelzpunkt: 120 °C (Zers.) Dichte: 1,91 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151033 CAS-Nummer: 13933-31-8 UN: 3077 ADR: 9,III |