Zinc phosphate hydrate, tech.
Lieferant: Thermo Fisher Scientific
11589.36
11589.A3
11589.36EA
72.6
EUR
11589.36
11589.A3
Zinc phosphate hydrate, tech.
Zinc phosphate hydrate
CAS No.: 7779-90-0
Molecular Formula: Zn3(PO4)2.xH2O
Formula Weight: 386.05(anhy)
Physical Form: -325 Mesh Powder
Solubility: Soluble in acetic acid, dilute mineral acids, ammonia. Insoluble in water
UN No.: UN3077
MDL No.: MFCD00036282
Formel:
O₈P₂Zn₃·xH₂O Molecular Weight: 386.08 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036282 CAS-Nummer: 7779-90-0 EINECS: 231-944-3 UN: 3077 ADR: 9 III Merck Index: 14,10151 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...